Learn More
Dräger™ Diffusion Tubes with Direct Indication
Catalog No. p-176713 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
2.5 to 188
Humidity Range:
1 to 16mg/L
Temperature Range (Metric):
0°C to 40°C
Catalog No. Measurement Range Humidity Range Temperature Range (Metric)
17-985-353C 2.5 to 188 1 to 16mg/L 0°C to 40°C
1 options
Catalog No. 17-985-353C Supplier Dräger™ Supplier No. 8101301
Only null left
Add to Cart
Add to Cart
Convenient, long-term TWA monitoring—direct reading, with no lab analysis required
Tubes sample ambient air by gaseous diffusion with no pumps or moving parts. To use, break off the tube end with the special holder, put the tube in the holder, and clip it to your pocket or lapel.
  • Obtain time-weighted average (TWA) measurements of exposure to toxic gases for periods of up to 8 hours
  • Reagent changes color as it reacts with the airborne contaminant
  • Final results are calculated by dividing the tube reading by the number of hours it was used
  • Designed and manufactured using ISO 90001:2000-certified processes
Order Info
Use with accessory clip-on holder (Cat. No. 17-986-4; available separately below). Ten tubes per pack.
† Higher concentration measurements possible with reduced sampling time.
Specifications
Components Measured Ammonia 20/a-d
Humidity Range 1 to 16mg/L
Measurement Range 2.5 to 188
Quantity 10
Response Time 8 hr.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C