Learn More
Dräger™ Flow Check Air Flow Indicator Kit: Smoke Generating Ampules
Catalog No. 19038776 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Draegar Flow Check Kit
Catalog No. For Use With (Equipment)
19-038-776 Draegar Flow Check Kit
1 options
Catalog No. 19-038-776 Supplier Dräger™ Supplier No. 6400812
  • $177.90
    $170.65 / Each of 1
    Save $7.25
    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Replacement ampules/nozzle for use with Dräger Flow Check Air Flow Indicator Kit
For Air Flow Indicator, see cat. no. 19038773
Specifications
For Use With (Equipment) Draegar Flow Check Kit
Type Smoke Generating Ampule