Learn More
Dräger™ Haz Mat Kits
Catalog No. 17985354R Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Type:
Hazmat Tube Kit
Catalog No. Type
17-985-354R Hazmat Tube Kit
1 options
Catalog No. 17-985-354R Supplier Dräger™ Supplier No. 4057185
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Accuro pump, 10 ft. (3m) extension hose for testing inaccessible areas, air current kit for determining wind direction and velocity, spare parts and tools for pump maintenance. Available with or without detector tubes.
Portable kit helps detect and measure a wide range of dangerous gases and vapors during initial assessments of potentially hazardous situations
  • Provides immediate onsite evaluations—important for quickly determining a course of action
  • Use included systematic sampling matrix with detector tubes (beginning with Polytest tube) to determine to which chemical group an unknown substance may belong
  • Also use the kit to quantitatively measure specific gases and vapors
  • Kit is housed in a durable, high-density polyethylene case with nonlocking latches and closed-cell foam insert that provides travel protection and organizes each component
Order Info
Kit measures 12 1/4L x 15 1/2W x 7 1/2 in.H (31.1 x 39.4 x 19.1cm). Net weight: 10 lb. (4.5kg).
Specifications
Type Hazmat Tube Kit
Includes Accuro pump, 10 ft. (3m) extension hose for testing inaccessible areas, air current kit for determining wind direction and velocity, spare parts and tools for pump maintenance. Available with or without detector tubes.