Learn More
Dräger™ Haz Mat Simultest Test Kit
Catalog No. 19500221 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Type:
Hazmat Simultaneous Test Kit w/o Pump
Catalog No. Type
19-500-221 Hazmat Simultaneous Test Kit w/o Pump
1 options
This item is not returnable. View return policy
No other type of tube should be substituted for use with these kits.
Catalog No. 19-500-221 Supplier Dräger™ Supplier No. 4056447
This item is not returnable. View return policy
No other type of tube should be substituted for use with these kits.
Includes
  • accuro™ pump
  • Pump spare parts kit
  • Air currents kit
  • 39 in. (1m) extension hose with adapter (includes scoring tool and manifold)
  • Simultaneous test set opener
  • Deluxe tube opener
  • Laminated instruction cards
  • Three each of Simultaneous Test Set† I (tests for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes)
  • Three each of Simultaneous Test Set† II (tests for sulfur dioxide, chlorine, hydrogen sulfide, carbon dioxide, and phosgene)
  • Four each of Simultaneous Test Set† III (tests for alcohols, aliphatic hydrocarbons, aromatics, chlorinated hydrocarbons, and ketones)
  • Hard-plastic carrying case
Simultaneously detects multiple contaminants
Provides Hazmat teams with onsite information about levels of inorganic/organic gases and vapors encountered during fires and afterward during salvage and overhaul operations.
  • Easy to use—just score and remove tube tips, insert special five-tube holder into manifold, and connect manifold to pump
  • After 10 complete pump strokes, tubes are ready for evaluation
Order Info
Individual test sets and adapter also sold separately.
†Each Simultest Set consists of five detector tubes in a rubber holder—ready for use. Each test set can be completed in one minute or less.
Specifications
Type Hazmat Simultaneous Test Kit w/o Pump
Includes Without Pump