Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Hydrogen Cyanide (HCN) XXS Sensor

Catalog No. 17980843 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Hydrogen Cyanide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-980-843 Hydrogen Cyanide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-980-843 Supplier Dräger™ Supplier No. 6810887
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XXS Replacement Sensor, for use with Pac 7000 and X-am 5000, X-am 5600 multi-gas detectors.

  • Measuring range: 0 to 50 ppm
  • Resolution: 0.1 ppm
  • Warranty: 2 years
  • Hydrogen Cyanide (HCN)

Specifications

Components Measured Hydrogen Cyanide
For Use With (Equipment) Pac 7000, X-am 5000, X-am 5600 Multigas Detectors
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.