Learn More
Dräger™ Multiwarn™ II Deluxe Multigas Monitors Accessories: Telescopic Sample Probe
Catalog No. 19087497 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
19-087497 Stainless Steel
1 options
Catalog No. 19-087497 Supplier Dräger™ Supplier No. 4510545
Add to Cart
Add to Cart
For use with Multiwarn II Deluxe Multigas Monitors
Telescopic Sample Probe
  • Stainless steel 1
  • 8 in. to 6.5 ft. (46cm to 2m)
Specifications
Type Telescopic Sample Probe
Length (English) 18 to 78 in.
Material Stainless Steel