Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ NiMH Battery Pack Kit for X-am™ 1000/2000/5000 Gas Monitors

Catalog No. 17985625 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
X-am 1000/2000/5000 Gas Detectors
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17-985-625 X-am 1000/2000/5000 Gas Detectors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-985-625 Supplier Dräger™ Supplier No. 8318785

Includes

Battery pack, charger cradle and single-power supply unit

Accessory for X-am 1000/2000/5000 Gas Detectors.

Accessory for X-am 1000/2000/5000 Gas Monitors.

Specifications

Color Black
For Use With (Equipment) X-am 1000/2000/5000 Gas Detectors
Type NiMH Battery Pack Kit
Includes Battery pack, charge cradle and single power supply
Voltage 110V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.