Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Pac™ III Sensors

Catalog No. 19815830 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide-HC (XS)
Diethylamine
Dimethyl Sulfide
Hydrogen Sulfide (XS)
Hydrogen Sulfide-HC (XS)
Nitric Oxide
Oxygen (XS)
Phosgene (COCL2)
Measurement Range:
0 to 100
0 to 1000
0 to 10000
0 to 25%
0 to 3
0 to 40
Warranty:
1 Year
3 Years
6 Months
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Measurement Range Warranty
19-815-830 Carbon Monoxide-HC (XS) 0 to 10000 1 Year
19-815-831 Oxygen (XS) 0 to 25% 3 Years
19-815-832 Hydrogen Sulfide-HC (XS) 0 to 1000 1 Year
19-815-829 Hydrogen Sulfide (XS) 0 to 100 3 Years
19-815-828 Phosgene (COCL2) 0 to 3 6 Months
19-815-836 Nitric Oxide 0 to 100 1 Year
19-815-833 Dimethyl Sulfide 0 to 40 1 Year
19-815-834 Diethylamine 0 to 100 1 Year
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-815-830 Supplier Dräger™ Supplier No. 6809120

Provide fast and accurate responses to hazardous gas levels

  • For use with Pac™ III single gas monitors

Specifications

Components Measured Carbon Monoxide-HC (XS)
Humidity Range 10 to 90%
Measurement Range 0 to 10000
Pressure Range 700 to 1300mbar
Resolution 5
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40° to +50°C
Warranty 1 Year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.