Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Reactive Gas Regulators

Catalog No. 19038749 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Flow Rate:
0.5 L/min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
19-038-749 0.5 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-038-749 Supplier Dräger™ Supplier No. 4594952
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compatible with Pac 7000 Single Gas Monitors

  • Constant flow
  • For Ammonia cylinder
  • With rotary valve
  • Flow rate: 0.5 LPM
  • Pressure: 1000psi
  • Cylinder connection: 5/8-18 UNF

Specifications

Connection Type 0.625 to 18 UNF
For Use With (Equipment) Pac III, Miniwarn, Multiwarn
Type Gas Regulator
Flow Rate 0.5 L/min.
For Use With (Application) Suitable for Ammonia Draeger calibration gas bottles
Includes Rotary valve
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.