Learn More
Dräger™ Short-Term Detector Tubes: Trichloroethylene
Catalog No. p-6467513 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
2 to 250
50 to 500
Tube Type:
Trichloroethylene 2/a2
Trichloroethylene 50/a
Response Time:
1.5 min.
8 to 4 min.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Response Time
19-810-245 50 to 500 Trichloroethylene 50/a 1.5 min.
17-985-311 2 to 250 Trichloroethylene 2/a2 8 to 4 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-810-245 Supplier Dräger™ Supplier No. 8101881
Only null left
Add to Cart
Add to Cart
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Trichloroethylene
Humidity Range 5 to 12mg/L
Measurement Range 50 to 500
No. of Strokes 5
Quantity 10
Response Time 1.5 min.
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15° to 40°C
Tube Type Trichloroethylene 50/a