Learn More
Dräger™ Short-Term Detector Tubes: Smoke Tube
Catalog No. 17985297 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Air Current
Catalog No. Components Measured
17-985-297 Air Current
1 options
Catalog No. 17-985-297 Supplier Dräger™ Supplier No. CH25301
  • $172.60
    $150.65 / Pack of 10
    Save $21.95
    13% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Description Detector Tubes
Components Measured Air Current
Quantity 10
Tube Type Air Current