Learn More
Dräger™ Short-Term Detector Tubes: Diethyl Ether
Catalog No. 17985218 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
100 to 4000
Catalog No. Measurement Range
17-985-218 100 to 4000
1 options
This item is not returnable. View return policy
Catalog No. 17-985-218 Supplier Dräger™ Supplier No. 6730501
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Diethyl Ether
Humidity Range 3 to 15mg/L
Measurement Range 100 to 4000
No. of Strokes 10
Quantity 10
Response Time 3 min.
Temperature Range (English) 59°F to 104°F
Temperature Range (Metric) 15° to 40°C
Tube Type Diethyl ether 100/a