Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Fluoride

Catalog No. 19046630 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.5 to 90ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range
19-046-630 0.5 to 90ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-046-630 Supplier Dräger™ Supplier No. 8103251
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides a reliable, cost-effective method for short-term, spot measurement of hydrogen fluoride gas concentrations.

The Dräger tube is a glass vial filled with a chemical reagent designed for the detection of hydrogen fluoride. It is used in conjunction with a Dräger gas detection pump, such as the Accuro™ pump, to measure gas concentrations through a colorimetric reaction.

  • Standard Measurement Range: 0.5 to 15 ppm
  • Measurement Time: Approximately 25 seconds
  • Number of Strokes (n): 10
  • Standard Deviation: 20% to 30%
  • Color Change: Blue-violet to yellow
  • Operating Temperature: 10°C to 40°C
  • Operating Humidity: 30% to 80%
  • Reaction Principle: HF + pH-indicator → yellow reaction product
  • Cross-Sensitivity: Mineral acids, such as hydrochloric acid or nitric acid, are also indicated. Basic gases, such as ammonia, can cause negative errors or prevent indication entirely.

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Hydrogen Fluoride 0.5/a
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range 30 to 80%
Measurement Range 0.5 to 90ppm
No. of Strokes 10/2
Quantity 10
Response Time 2 min./25 sec.
Temperature Range (English) 50°F to 104°F
Temperature Range (Metric) 10° to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.