Learn More
Dräger™ Short-Term Detector Tubes - De Tube Natural Gas Test
Shop All Draeger Safety, Inc. ProductsDescription
Dräger Tubes are glass vials containing chemical reagents that change color to indicate the measured concentration of target chemicals.
- Compatibility: Dräger Accuro gas detection pump, Accuro 2000, and Quantimeter 1000.
- Applications: Determination of concentration peaks, measurement of inhalation area exposures, detection of potential leakages, and analysis of air in confined spaces like sewers, shafts, or tanks.
- Operating Principle: A calibrated 100 mL sample of air is drawn through the tube, triggering a color change proportional to the chemical concentration.
- Handling and Storage: Stable under normal conditions; store at 25°C (77°F) or lower.
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Specifications
| Components Measured | Natural Gas |
| For Use With (Equipment) | Accuro Drager Gas Detector Pump |
| Type | Gas Detection Tube |
| Humidity Range | 40mg/L |
| Measurement Range | Qualitative |
| No. of Strokes | 2 |
| Quantity | 10 |
| Response Time | 40 sec. |
| Temperature Range (English) | 32°F to 122°F |
| Temperature Range (Metric) | 0°C to 50°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.