Learn More
Description
Dräger short-term detector tubes for the measurement of hydrazine gas.
- Product: Dräger tube Hydrazine 0.01/a
- Measurement Principle: Chemical reagent in a glass vial reacts to the target substance, causing a color change proportional to the concentration.
- Usage: Designed for use with the Dräger Accuro™ gas detection pump, which draws a 100 mL air sample through the tube.
- Application: Suitable for determining concentration peaks of hydrazine.
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Specifications
| Components Measured | Hydrazine 0.01/a |
| For Use With (Equipment) | Accuro Drager Gas Detector Pump |
| Type | Gas Detection Tube |
| Measurement Range | 0.01 to 6ppm |
| No. of Strokes | 5 |
| Quantity | 10 |
| Response Time | 20 to 30/1 min. |
| Temperature Range (English) | 32°F to 86°F |
| Temperature Range (Metric) | 0°C to 30°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.