Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes - Nitric Acid

Catalog No. 17985268 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
1 to 50ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range
17-985-268 1 to 50ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-985-268 Supplier Dräger™ Supplier No. 6728311
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reliable and cost-effective gas detection tubes for spot measurements of nitric acid in various industrial applications.

Dräger -Tubes™ are glass vials containing a chemical reagent that changes color in the presence of specific gases, allowing for direct concentration readings.

  • Gas: Nitric Acid
  • Measurement Range: 1 to 15 ppm (standard), 5 to 50 ppm
  • Measurement Time: 5 seconds to 15 minutes
  • Compatibility: Used with Dräger Accuro™ bellows pump, Accuro 2000, or Quantimeter 1000
  • Applications: Determination of concentration peaks, exposure measurement, leakage detection, and air analysis in confined spaces

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Nitric Acid 1/a
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range 3 to 15mg/L
Measurement Range 1 to 50ppm
No. of Strokes 10/20
Quantity 10
Response Time 2 to 4 min.
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5° to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.