Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes - Oil

Catalog No. 19810246 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
5mg/m3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range
19-810-246 5mg/m3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-810-246 Supplier Dräger™ Supplier No. 8103111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For measurement of oil in air.

  • For use with Aerotest Navy system, that is designed for special demands of military applications
  • Measuring Range: 5mg/m3
  • No. of Detector Tubes: 10

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Oil
For Use With (Equipment) Aerotest Navy System
Type Gas Detection Tube
Measurement Range 5mg/m3
Quantity 10/Pk.
Tube Type Short-term
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.