Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes - Petroleum Hydrocarbons

Catalog No. 17985286 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
100 to 2500ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range
17-985-286 100 to 2500ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-985-286 Supplier Dräger™ Supplier No. 6730201
  • $210.00
    $177.65 / Pack of 10

    Save $32.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Short-term detector tube for cost-effective and reliable measurement of petroleum hydrocarbons in gas.

DrägerTubes are glass vials filled with a chemical reagent for the measurement of gas. They are designed for manual or automatic use with Dräger gas detection pumps. Applications include determination of concentration peaks, measurement of exposures in the inhalation area, identifying leakages, and analysis of air in confined spaces.

  • Target: Petroleum hydrocarbons
  • Standard range: 100 to 2,500 ppm (at 20°C, 1013 hPa)
  • Measurement time: 30 sec

  1. Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
  2. Safety Equipment Institute (SEI) certified.

Specifications

Components Measured Petroleum Hydrocarbons 100/a (Acetylene)
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range Less than30mg/L
Measurement Range 100 to 2500ppm
No. of Strokes 2
Quantity 10
Response Time 30 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.