Learn More
Description
DrägerTubes are glass vials filled with a chemical reagent designed for gas detection. When a 100 mL sample of air is drawn through the tube, the reagent changes color to indicate the presence and concentration of the target substance.
- Product Type: Short-term tubes
- Measurement Time: 10 sec to 15 min
- Configuration: Tubes with one indicating layer
- Compatibility: Designed for use with Dräger Accuro™ gas detection pump, Accuro 2000, or Quantimeter 1000
- Applications: Determination of concentration peaks, measurement of exposures in the inhalation area, leak detection, and analysis of air in sewers, shafts, tanks, or other confined spaces
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Specifications
| Components Measured | Phenol 1/b |
| For Use With (Equipment) | Accuro Drager Gas Detector Pump |
| Type | Gas Detection Tube |
| Humidity Range | 1 to 18mg/L |
| Measurement Range | 1 to 20ppm |
| No. of Strokes | 20 |
| Quantity | 10 |
| Response Time | 5 min. |
| Temperature Range (English) | 50°F to 86°F |
| Temperature Range (Metric) | 10°C to 30°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.