Learn More
Description
Dräger -Tubes are glass vials designed for the detection of specific gases and vapors.
- Product: Phosphoric Acid Ester 0.05/a
- Standard measurement range: 0.05 ppm (at 20°C, 1013 hPa)
- Measurement time: 5 sec.
- Sample volume: 100 mL
- Operation: Utilizes the Dräger Accuro™ bellows pump or automatic pumps such as the Accuro 2000 and Quantimeter 1000.
- Principle: Targeted chemicals cause a color change within the reagent, with the length of the discoloration indicating the concentration.
- Applications: Determination of concentration peaks, exposure measurement in inhalation areas, leakage detection, and analysis of air in confined spaces like sewers, shafts, and tanks.
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Specifications
| Components Measured | Phosphoric Acid Esters 0.05/a |
| For Use With (Equipment) | Accuro Drager Gas Detector Pump |
| Type | Gas Detection Tube |
| Humidity Range | 3 to 18mg/L |
| Measurement Range | Up to 0.05ppm |
| No. of Strokes | 10 |
| Quantity | 10 |
| Response Time | 5 min. |
| Temperature Range (English) | 50°F to 104°F |
| Temperature Range (Metric) | 10° to 40°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.