Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes: Water Vapor

Catalog No. 17985321D Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.05 to 1
1 to 40
20 to 1500
20 to 40mg/L
3 to 60
5 to 200
Tube Type:
Water vapor 0.1
Water vapor 0.1/a
Water vapor 1/b
Water vapor 20/a-P
Water vapor 3/a
Water vapor 5/a-p
Response Time:
1.5 min.
10 to 5 min.
2 min.
20/40 sec.
25 min.
Not Applicable
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Response Time
17-985-321D 5 to 200 Water vapor 5/a-p 25 min.
18-556N 3 to 60 Water vapor 3/a Not Applicable
17-985-317 0.05 to 1 Water vapor 0.1/a 1.5 min.
18-556Q 20 to 40mg/L Water vapor 1/b 20/40 sec.
19-046-633 20 to 1500 Water vapor 20/a-P 10 to 5 min.
17-985-651 1 to 40 Water vapor 0.1 2 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-985-321D Supplier Dräger™ Supplier No. 6728531
Add to Cart
Add to Cart

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ Customer Service for more information.

Specifications

Components Measured Water Vapor
Humidity Range Not Applicable
Measurement Range 5 to 200
Quantity 10
Response Time 25 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Tube Type Water vapor 5/a-p
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.