Learn More
Dräger™ X-am™ 3000 Gas Monitor: Calibration Gas Cylinder, 50p
Catalog No. 19046618 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
X-am 3000 Gas Monitor
Catalog No. For Use With (Equipment)
19-046-618 X-am 3000 Gas Monitor
1 options
Catalog No. 19-046-618 Supplier Dräger™ Supplier No. 4594639
  • $557.00
    $413.65 / Each of 1
    Save $143.35
    26% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Calibration Gas Cylinder for X-am 3000 gas monitors
For use with X-am 3000 gas monitors
50p
Specifications
For Use With (Equipment) X-am 3000 Gas Monitor
Type Calibration Gas Cylinder
Includes 50% LEL CH4, 100ppm Carbon Monoxide, 25ppm H2S, 17% O2