Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ X-plore™ 3500 Half-Mask Respirators

Catalog No. 19088346 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-088-346 Large
19-088-345 Small
19-088-344 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-088-346 Supplier Dräger™ Supplier No. R55352
This item is not returnable. View return policy

Made of soft and skin-friendly DrägerFlex material, X-plore™ 3500 features a drop-down strap design that enables the mask to be removed simply by releasing the fastener at the neck.

  • Modern, low-profile mask body design
  • Extended field of vision and room to easily use visors and shields
  • Can be worn comfortably under helmets and visors
  • Soft and skin-friendly DrägerFlex material
  • Soft, lightweight and flexible strap system
  • Flexi-Fit head harness conforms to the contours of head without pressure or hair entanglement
  • Innovative X-Guided cross-over strap system ensure even weight distribution and an even seal
  • 2-point bayonet connector with markings for insertion of the filter
  • Flexible nose seal fits securely with safety glasses and goggles
  • Drop-down strap design enables mask to be removed simply by releasing the fastener at the neck
  • Easy to disassemble, clean and reassemble

Specifications

Certifications/Compliance AS NZS 1716
Certifications (EN) EN 140
Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Blue
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Large
For Use With (Equipment) X-plore™ Bayonet Filters and Cartridges
Includes Drop-down harness system, Reasealable storage bag
Model R55352
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.