Learn More
Enzo Life Sciences DR2313 (25mg). CAS: 284028-90-6
SDP
Supplier:  Enzo Life Sciences ALX270452M025
Encompass_Preferred
Potent, water soluble competitive PARP inhibitor (IC50=0.20µM and 0.24µM for PARP-1 and PARP-2 respectively). Alternative name: 2-Methyl-3,5,7,8-tetrahydrothiopyrano[4,3-d]pyrimidine-4-one. Purity: ≥98% (HPLC (UV)). Solubility: Soluble in DMSO, methanol or water. Long Term Storage: -20°C.
Catalog No. 50-200-8575
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.