Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Short-Term Detector Tubes: Ammonia

Catalog No. 17985177 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
0.25 to 3
0.5 to 10%
2 to 30
5 to 100
5 to 700
Tube Type:
Ammonia 0.25/a
Ammonia 0.5%/a
Ammonia 2/a
Ammonia 5/a
Ammonia 5/b
Humidity Range:
3 to 12mg/L
Less than20mg/L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Tube Type Humidity Range
17-985-177 2 to 30 Ammonia 2/a Less than20mg/L
18-556A 0.25 to 3 Ammonia 0.25/a Less than20mg/L
19-038-830 5 to 100 Ammonia 5/b Less than20mg/L
17-985-178 5 to 700 Ammonia 5/a Less than20mg/L
17-985-179 0.5 to 10% Ammonia 0.5%/a 3 to 12mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-985-177 Supplier Dräger™ Supplier No. 6733231
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Detector tubes

Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.

Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.

Order Info

Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.

Specifications

Description Detector Tubes
Components Measured Ammonia
For Use With (Equipment) Accuro Drager Gas Detector Pump
Type Gas Detection Tube
Humidity Range Less than20mg/L
Measurement Range 2 to 30
No. of Strokes 5
Quantity 10
Response Time 1 min.
Temperature Range (English) 50°F to 122°F
Temperature Range (Metric) 10° to 50°C
Tube Type Ammonia 2/a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.