Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Drain Cover Carrying Storage Case
SDP

Catalog No. 18366633 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polyethylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-366-633 Polyethylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-633 Supplier PIG™ Supplier No. PLR286
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hard-shell carrying case for DrainBlocker Drain Cover.

  • Threaded, screw-in brackets mount on any vertical surface
  • Rigid polyethylene shell is corrosion and UV resistant to protect your drain cover indoors or out
  • High-visibility yellow is easy to spot during an emergency
  • Large label makes identification fast and accurate
  • Pull-off end cap with twist lock closure for quick access, attached to case to prevent loss
  • Textured rubber handle makes case easy to carry
  • Drain cover not included
  • For square 30 to 36 in., rectangular 36 x 60 in., or round up to 30 in. diameter drain covers
  • 7.5 x 43 in.

Specifications

Color Yellow
Material Polyethylene
Type Drain Cover Carrying Case
Dimensions (L x W) 43 x 7.5 in.
Weight (English) 8.94 lb.
Weight (Metric) 4.1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.