Learn More
Description
- Frequently used with PCTE (Polycarbonate) and PETE (Polyester) membranes to increase flow
- Spacer between stacked membranes
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Membrane Filter |
| Format | Disc |
| For Use With (Equipment) | Filtration Systems |
| Material | Polyester |
| Material (Membrane) | Polyester |
| Sterility | Non-sterile |
| Type | Membrane Filter |
| Diameter (Metric) | 13 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.