Learn More
New Pig DrainBlocker™ Drain Cover
SDP
Catalog No. 18389145 Shop All New Pig Corporation Products
Change view
Click to view available options
Color:
Yellow
Catalog No. Color
18-389-145 Yellow
1 options
This item is not returnable. View return policy
Catalog No. 18-389-145 Supplier New Pig Supplier No. PLR404
Add to Cart
Add to Cart
This item is not returnable. View return policy
Prevent spills from going down your storm drains with the patented design that seals tight and won't rip.
  • Durable, long-lasting design withstands long-term outdoor use Wipes clean so it can be reused
  • Tougher than your standard drain covers! PIG-tough vinyl top resists stretching, ripping and tearing
  • Super-sealing bottom layer conforms to uneven surfaces for a tight seal
  • Withstands prolonged exposure to diesel fuel and gasoline
  • Use for quick spill response or leave it down for long-term drain protection where liquid transfers, washdowns or other leak- and spill- prone jobs are common
  • DuPont Elvaloy technology provides lifelong flexibility plus chemical, UV and infrared light resistance to vinyl top
  • High-visibility yellow top layer draws attention to spills Includes a nylon storage bag for easy transportation
Specifications
Color Yellow