Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DRAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119877 Supplier Invitrogen™ Supplier No. PA5119877
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: SW1990 cell lysate, Jurkat cell lysate, SK-Br-3 cell lysate, mouse testis tissue lysate, rat testis tissue lysate, rat testis tissue, human cervix tissue. Subcellular Location: Nucleus.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DRAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene DRAP1
Gene Accession No. A0JPP1, Q14919, Q9D6N5
Gene Alias 2310074H19Rik; DR1 associated protein 1; Dr1 associated protein 1 (negative cofactor 2 alpha); dr1-associated corepressor; Dr1-associated protein 1; DR1-associated protein 1 (negative cofactor 2 alpha); DRAP1; drap1 {ECO:0000250; NC2-alpha; negative cofactor 2 alpha; negative co-factor 2-alpha; negative cofactor 2-alpha; UniProtKB:Q14919}
Gene Symbols DRAP1
Host Species Rabbit
Immunogen Synthetic peptide within human NC2 alpha aa 150-200.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10589, 293674, 66556
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.