Learn More
Med Vet International Sheet Drape 53X77 Sterile 20 Ea/Case
Supplier:  Med Vet International DYNJP2414
Encompass_Preferred
Medline's surgical drapes have received an upgrade! Competitive with the new industry standard, a high-quality SMS material provides exceptional barrier protection (double the hydrostatic pressure of Eclipse fabric) without sacrificing comfort or softness. Our SMS fabric binds three layers of fabric--a densely-packed meltblown layer sandwiched between a strong spunbond outer layers--to provide incredible fluid repellency. Progressive Reicofil technology offers customers increased fabric uniformity and drapability, equipping you with the market's latest-generation of SMS. You really can't get more from a surgical drape. Three-Quarter Drape 53"x77". DROP SHIP ITEM
Catalog No. 50-209-1798
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.