Learn More
Med Vet International Disposable Surgical Veterinary Drape SMS 39"" X 100 Yards, White, 1 roll
Supplier:  Med Vet International SMS39W
Encompass_Preferred
This high quality surgical drape is made of SMS non-woven material. Consists of two white outer layers of SMS and a middle layer of spunbound polypropylene. Our disposable vet drape can surround the surgical area with barrier proof material during veterinary surgery. Can also be used as a drape to control and contain fluid, as fenestrated material or as a sterilization wrap. Non-sterile. Autoclavable Fabric is low lint, helping to reduce wound contamination. Microfibers filter out dry particles and many water-based liquids. First-rate option for a variety of innovative surgical draping solutions and also for general medical applications. Cloth-like feel Autoclavable
Catalog No. 50-209-1835
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.