Learn More
Specifications
Specifications
| For Use With (Application) | Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy |
| Content And Storage | -20°C |
| Product Line | Novus |
| Quantity | 200 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
