Learn More
Cayman Chemical DRB, 53-85-0, 10 mg
SDP
Supplier:  Cayman Chemical 1001030210
Encompass_Preferred
Inhibits CKII (IC50 range of 4-10 µM), Cdk7 (IC50 = ~20 µM), Cdk8 (IC50 = ~20 µM), and Cdk9 (IC50 = 3 µM); inhibits elongation during RNA polymerase II transcription; triggers p53-dependent apoptosis of human colon adenocarcinoma cells; inhibits trans-activated transcription of the Tat protein of HIV-1 (IC50 = ~4 µM)
Catalog No. NC1379794
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.