Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589944 Supplier Thermo Scientific Supplier No. PA589944
Only null left
Add to Cart
Add to Cart

DRD3 Polyclonal Antibody

DRD3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DRD3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DRD3
Gene Accession No. P19020, P30728
Gene Alias D(3) dopamine receptor; D3 receptor; D3DR; D3nf; D3R; Dopamine 3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopamine receptor D3; dopaminergic receptor D3; Drd3; essential tremor 1; ETM1; FET1
Gene Symbols DRD3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Dopamine Receptor D3 (Dopamine Receptor D3 (DRD3)) (NP_0007872).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13490, 29238
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.