Learn More
Abcam DREADD Agonist 21, DREADD agonist, 1MG
Supplier:  Abcam AB2355451MG
Encompass_Preferred
MW 278.35 Da, Purity >98%. Potent, selective muscarinic hM3Dq DREADD (Designer Receptors Exclusively Activated by Designer Drugs) agonist (EC₅₀ = 1.7 nM). Highly selective for hM₃Dq over endogenous hM₃ receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6064
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.