Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DreamTaq™ Buffers (10X)

Catalog No. FERB71 Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Colorless
Green
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
FERB71 Green
FERB65 Colorless
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERB71 Supplier Thermo Scientific™ Supplier No. B71
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific 10X DreamTaq Buffer is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase.

Thermo Scientific DreamTaq Buffer (10X), available in colorless and green formats, is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase. It contains both KCl and (NH4)2SO4 to provide high specificity of primer annealing at a wide range of MgCl2 concentrations. 10X DreamTaq buffer includes 20 mM MgCl2

In addition, 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes. The density reagent allows direct loading of PCR products on a gel. The blue dye (migrates with 3–5 kb DNA fragments in 1% agarose gel) and the yellow dye (migrates faster than 10 bp DNA fragments in 1% agarose gel) are included for monitoring electrophoresis progress. The dyes have excitation peaks at 424 nm and 615 nm.

Notes

  • For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless DreamTaq Buffer.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DreamTaq Green Buffer
Color Green
Concentration 10X
For Use With (Application) Hot-start PCR
Quantity 4 x 1.25 mL
Formulation Nonidet P40 Ethyl phenolpoly(ethyleneglycolether) 9016-45-9 ( <0.9 ), Polyethylene glycol 25322-68-3 ( 7-13 ), Sucrose 57-50-1 ( 10-30 )
Includes • 10X DreamTaq Green Buffer (with 20 mM MgCl2), 4 x 1.25 mL

Store in a non-frost-free freezer at -20°C.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.