Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drebrin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7087841 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6158 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6158 Supplier Proteintech Group Inc Supplier No. 102601AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The cortical cytoskeleton, a highly cross-linked submembranal F-actin filament network, plays an important role in cell proliferation, differentiation, migration, cell morphology and oncogenic transformation. Drebrin (developmentally-regulated brain protein) is a major neuronal F-actin binding protein involved in the control of actin dynamics and neuronal morphogenesis. Three major isoforms of drebrin, resulting from alternative splicing of a single gene, have been identified in the brain: the embryonic isoforms designated E1 and E2 and the adult or A isoform (95-120 kDa).

Specifications

Antigen Drebrin
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DBN1
Gene Accession No. Q07266, Q16643, Q9QXS6
Gene Alias D0S117E, DBN1, Drebrin, drebrin 1
Gene Symbols Dbn1
Host Species Rabbit
Immunogen Drebrin Fusion Protein Ag0371
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1627, 56320, 81653
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.