Learn More
New Pig™ Drench Shower Tester
SDP
Catalog No. 18389155 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Drench Shower Tester
Catalog No.
18-389-155 Type: Drench Shower Tester
1 options
This item is not returnable. View return policy
Catalog No. 18-389-155 Supplier New Pig™ Supplier No. TLS609
Add to Cart
Add to Cart
This item is not returnable. View return policy
Before you run your weekly ANSI drench shower test, install this clear vinyl funnel to channel the water away.
  • ANSI standards require drench showers to be tested weekly, which can leave your floors (and workers) all wet
  • Tester catches water and channels it to a floor drain or container 12 in. diameter funnel is made of clear vinyl with reinforced top opening
  • Durable material keeps its shape and allows easy visual inspection
  • Attached 25' flat vinyl hose has 2 in. diameter to drain up to 20 gallons per minute
  • Includes stainless steel mounting bracket and four S-type hooks; installs easily on most dome-style drench showers
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov