Learn More
Med Vet International Fluoroscope Cover 3M Steri-Drape 35 X 43 inch Fluoroscopes, 1012, 40/Case
Supplier:  Med Vet International 5715CS
Encompass_Preferred
This sterile 3M surgical drape forms a protective barrier to prevent potential contamination It's made of strong polyethylene to withstand equipment manipulation while in use The sturdy plastic material resist accidental penetration of fluids A clear, see-through design enables a full view of the fluoroscope The drape provides a quick fit for easy setup An adhesive closure style keeps the drape securely in place It's disposable to ease cleanup 35 inches wide and 43 inches long
Catalog No. 50-283-1213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.