Learn More
Med Vet International Orthopedic Padding Adhesive 3M Reston 7.875 X 11.75 inch Foam Nonsterile, 1560M, Each
Supplier:  Med Vet International 5788EA
Encompass_Preferred
Self-adhering foam designed to distribute body weight to help prevent skin breakdown and pressure ulcers, and to prevent skin damage from splints, casts and prostheses 7/16-inch thick Medium support pad can be cut, layered and shaped to be adapted for many applications Reston self-adhering foam products should not be placed in direct contact with an open wound 7-7/8 x 11¾ inches
Catalog No. 50-283-0821
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.