Learn More
Thermo Scientific™ DRI™ Amphetamine Drugs of Abuse Assays
Description
Eliminate time-consuming steps while achieving high analytical accuracy when you use Thermo Scientific™ DRI™ Amphetamine Drugs of Abuse Assays—ready to use in liquid form. These assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost-control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | MGC Multi-Drug Controls |
| Description | DRI Amphetamine Drugs of Abuse Assays |
| Detectable Analytes | Amphetamine (AMP), Methamphetamine (mAMP) |
| DoA Calibrators | Multi-drug Calibrator |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 1 x 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.