Learn More
Medchemexpress LLC Dri-C21045 | 2101765-81-3 | 98.1% | 100 MG

Supplier: Medchemexpress LLC HY120323100MG
DRI-C21045 is a small-molecule inhibitor of the CD40-CD40L protein-protein interaction, supplied for research use with reported in vitro potency (IC50 ≈ 0.17 μM) and high purity suitable for biochemical and cell-based studies.
- Potent inhibitor of the CD40-CD40L protein-protein interaction (reported IC50 ~0.17 μM).
- High purity suitable for research (≈98.1%).
- Molecular formula C32H24N2O7S and molecular weight 580.61 g/mol.
- Available in small research pack sizes for in vitro studies.
- Intended for biochemical and cell-based research; not for therapeutic use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.