Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-(4'-(4-(methoxycarbonyl)benzamido)biphenyl-4-ylcarboxamido)naphthalene-1-sulfonic acid | 2101765-81-3 | 98.1% | 10 MG
SDP

Supplier:  Medchemexpress LLC HY12032310MG

Encompass_Preferred

DRI-C21045 is a small-molecule inhibitor of the CD40-CD40L protein-protein interaction used in research to modulate immune costimulatory signaling. It is reported to be active at submicromolar concentrations (IC50 ≈ 0.17 μM) and is supplied as a solid research compound.

  • Potent CD40-CD40L interaction inhibitor (IC50 ≈ 0.17 μM).
  • High chemical purity suitable for research applications.
  • Molecular weight 580.61 and formula C32H24N2O7S.
  • Off-white to gray solid, typically supplied in 10 mg vials.
  • Soluble in DMSO; handle per standard laboratory safety procedures.

Catalog No. 50-002-34424


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.