Learn More
Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls
Description
Reduce time-consuming reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls—ready-to-use controls in lilquid form. Delivering high analytical accuracy, these assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
Specifications
| Description | DRI Cotinine Drugs of Abuse Controls |
| Concentration | 700 ng/mL |
| Detectable Analytes | Creatinine |
| Sample Type | Urine |
| Type | Control |
| Level | High (700ng/mL) and Low (300ng/mL) |
| Quantity | 1 x 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.