Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamedix™ Dri-Dye Check Strips
SDP

Catalog No. 23451050
Encompass
Change view
Click to view available options
Wavelength:
405 nm
450 nm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength
23-451-050 405 nm
23-451-051 450 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-451-050 Supplier Diamedix™ Supplier No. 790900
Add to Cart
Add to Cart

Includes

6 pre-filled strips per kit, re-usable strip tray (included for use with plate readers), linearity plots, calibration and repeatability charts, instructions

For performance verification of microstrip and microplate readers. Diamedix Dri-Dye Check Strips provide easy and economical way to monitor calibration, linearity and repeatability of microstrip and plate readers.

Dri-Dye™ Check Strips

  • Each kit comes with Calibration charts, linearity plots and repeatability records, six prefilled strips and a reusable strip tray for use with plate readers
  • Strips simulate true test conditions better than neutral density filters by providing information about filtering capacity
  • Simply add water, read and compare results to pre-determined acceptance criteria
  • Long shelf life at room temperature
  • NIST traceable absorbance calibration
  • Wavelengths available 405, 450 and 492nm

Specifications

Type Check Strips
For Use With (Application) For performance verification of microstrip and microplate readers
Analytical Method ELISA
Certifications/Compliance NIST Traceable
Wavelength 405 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.