Learn More
Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets
Description
Streamline your testing process while maintaining high analytical accuracy by utilizing Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets along with our Thermo Scientific DRI Ethyl Glucuronide (EtG) Drugs of Abuse Assays and Calibrators. These control sets are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
- Specific detection of Ethyl Glucuronide (EtG)
- Qualitative or semi-quantitative modes
- Excellent correlation to LC-MS/MS
- Applications for an array of clinical chemistry analyzers
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | DRI Ethyl Glucuronide (EtG) 750 Control. For Criminal Justice and Forensic Use Only. For sale in the U.S.A. only. |
| Concentration | 750 ng/mL |
| Detectable Analytes | Ethyl Glucuronide (EtG) |
| Sample Type | Urine |
| Type | Control |
| Quantity | 1 x 25 mL |
For sale in the U.S.A. only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.