Learn More
Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Assay

Description
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Assays. Field-tested on a wide range of clinical analyzers, including the Thermo Scientific™ Indiko™ Clinical and Specialty Chemistry System, these liquid, ready-to-use assays have been widely accepted in laboratories where cost control and optimal productivity are key issues. These assays are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
- Specific detection of Ethyl Glucuronide (EtG)
- Qualitative or semi-quantitative modes
- Excellent correlation to LC-MS/MS
- Applications for an array of clinical chemistry analyzers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | DRI Ethyl Glucuronide (EtG) Control Sets |
| Description | DRI Ethyl Glucuronide (EtG) Drugs of Abuse Assay. Criminal Justice and Forensic use only. For sale in the U.S.A. only. |
| Detectable Analytes | Ethyl Glucuronide (EtG) |
| DoA Calibrators | Ethyl Glucuronide CE Calibrator |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 1 x 68 mL |
For sale in the U.S.A. only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.