Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Calibrators
GSA_VA

Catalog No. 23278366
Change view
Click to view available options
Concentration:
0 ng/mL
100 ng/mL
1000 ng/mL
2000 ng/mL
500 ng/mL
Quantity:
1 x 10 mL
1 x 25 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration Quantity
23-278-366 1000 ng/mL 1 x 10 mL
23-278-360 0 ng/mL 1 x 25 mL
23-278-361 100 ng/mL 1 x 10 mL
23-278-363 500 ng/mL 1 x 10 mL
23-278-368 2000 ng/mL 1 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
For Criminal Justice and Forensic use only.
Catalog No. 23-278-366 Supplier Thermo Scientific™ Supplier No. 10015938
Add to Cart
Add to Cart
This item is not returnable. View return policy
For Criminal Justice and Forensic use only.

Save time while maintaining accuracy with these liquid, ready-to-use calibrators, which are for Criminal Justice and Forensic Use Only and are for sale in U.S.A. only.

Ensure accuracy and precision of your drug testing results by partnering these liquid, ready-to-use Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Calibrators with our Thermo Scientific DRI Ethyl Glucuronide (EtG) Drugs of Abuse Assay and Control Sets. These calibrators are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.

  • Specific detection of Ethyl Glucuronide (EtG)
  • Qualitative or semi-quantitative modes
  • Excellent correlation to LC-MS/MS
  • Applications for an array of clinical chemistry analyzers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Ethyl Glucuronide (EtG) 1000 Calibrator. For Criminal Justice and Forensic Use Only. For sale in the U.S.A. only.
Concentration 1000 ng/mL
Detectable Analytes Ethyl Glucuronide (EtG)
Sample Type Urine
Type Calibrator
Packaging 1 Bottle
Quantity 1 x 10 mL

For sale in the U.S.A. only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.