Learn More
Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Assay
Description
Utilize the globally valued accuracy, precision, lot-to-lot dependability and stable shelf life of the Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Assay, which offers applications for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing and developing new assays to respond to ever-emerging drug screening needs. This assay is for sale in the U.S.A. and Canada only. Supports new SAMHSA guidelines (effective October 1, 2017)
- Specific detection of hydrocodone
- Qualitative or semi-quantitative modes
- Excellent correlation to LC-MS/MS
- Applications for an array of clinical chemistry analyzers
- For sale in the U.S.A. and Canada only
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | DRI Hydrocodone Control Set |
| Description | DRI Hydrocodone Drugs of Abuse Assay |
| Detectable Analytes | Hydrocodone |
| DoA Calibrators | Hydrocodone Calibrator, Negative Urine Calibrator |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 1 x 500 mL |
Disclaimers: Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.