Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators

Catalog No. 23278378
Change view
Click to view available options
Concentration:
100 ng/mL
1000 ng/mL
300 ng/mL
500 ng/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
23-278-378 300 ng/mL
23-278-377 100 ng/mL
23-278-379 500 ng/mL
23-278-380 1000 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 23-278-378 Supplier Thermo Scientific™ Supplier No. 10018080
Add to Cart
Add to Cart
This item is not returnable. View return policy

Save time while maintaining accuracy with the liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators.

Ensure accuracy and precision of your drug testing results by partnering these liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators with our Thermo Scientific DRI Hydrocodone Drugs of Abuse Assay and Control Set.These calibrators are for sale in the U.S.A. and Canada only.

  • Specific detection of hydrocodone
  • Qualitative or semi-quantitative modes
  • For sale in the U.S.A. and Canada only



TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Hydrocodone Drugs of Abuse Calibrator
Concentration 300 ng/mL
Detectable Analytes Hydrocodone
Sample Type Urine
Type Calibrator
Packaging 1 Bottle
Quantity 1 x 10 mL

Disclaimers: Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.